siRNA Details
Gene Symbol: TMSB4X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021109.3 | NP_066932.1 | 1 | 190 | ||
GenBank mRNA | AK055976.1 | - | 2 | 1264 | ||
AK057827.1 | - | 3 | 187 | |||
AK311747.1 | - | 3 | 188 | |||
AV715954.1 | - | 3 | 190 | |||
BC001631.1 | - | 1 | 171 | |||
BC016732.1 | - | 1 | 191 | |||
BC022857.1 | - | 1 | 191 | |||
BC061586.1 | - | 3 | 177 | |||
BC062436.1 | - | 1 | 191 | |||
BC064954.1 | - | 3 | 171 | |||
BC070248.1 | - | 3 | 188 | |||
BC070249.1 | - | 1 | 187 | |||
BC083509.1 | - | 1 | 187 | |||
BC092437.1 | - | 1 | 187 | |||
BC101790.1 | - | 3 | 174 | 8069296 | ||
BC101792.1 | - | 3 | 174 | 8069298 | ||
BC104464.1 | - | 1 | 141 | 40031330 | ||
BC104465.1 | - | 1 | 141 | 40031337 | ||
BC104666.1 | - | 3 | 191 | |||
BC107789.1 | - | 3 | 194 | |||
BC128102.1 | - | 3 | 159 | 40112039 | ||
BC128103.1 | - | 3 | 162 | 40112043 | ||
BC139925.1 | - | 1 | 176 | 40148985 | ||
BC141975.1 | - | 3 | 162 | 40112044 | ||
BC141976.1 | - | 3 | 162 | 40112045 | ||
BC151215.1 | - | 1 | 177 | 40148015 | ||
BT007090.1 | - | 2 | 112 | |||
M17733.1 | - | 3 | 189 | |||
X02493.1 | - | 2 | 92 |
Gene Symbol: TMSB4XP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC090885.1 | - | 1 | 51 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
