siRNA Details
Gene Symbol: SNX29P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002939.3 | 7, 8 | 960 | |||
GenBank mRNA | AK023827.1 | - | 8, 9 | 1183 | ||
BX648280.1 | - | 9, 10 | 1632 |
Gene Symbol: SNX29P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045011.1 | 4, 5 | 357 | |||
GenBank mRNA | BC041583.1 | - | 1, 2 | 221 | 4765274 | |
DR003051.1 | - | 4, 5 | 363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
