siRNA Details
Gene Symbol: MTURN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152793.2 | NP_690006.2 | 2 | 470 | ||
XM_005249652.3 | XP_005249709.1 | 2 | 413 | |||
GenBank mRNA | AK096533.1 | - | 2 | 413 | ||
AK294982.1 | - | 2 | 200 | |||
AL834391.1 | - | 2 | 404 | |||
AY054121.1 | - | 2 | 266 | |||
BC062423.1 | - | 2 | 221 | |||
BI560774.1 | - | 2 | 474 | |||
DT217816.1 | - | 2 | 405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
