siRNA Details
Gene Symbol: CCZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015622.5 | NP_056437.4 | 4, 5 | 482 | ||
GenBank mRNA | AF151801.1 | - | 4, 5 | 526 | ||
BC067810.1 | - | 4, 5 | 492 | |||
BC132749.1 | - | 4, 5 | 467 | 40146771 | ||
BC132751.1 | - | 4, 5 | 467 | 40146773 | ||
BI546694.1 | - | 4, 5 | 474 |
Gene Symbol: CCZ1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198097.3 | NP_932765.1 | 4, 5 | 490 | ||
GenBank mRNA | AK292326.1 | - | 4, 5 | 458 | ||
AK296026.1 | - | 3, 4 | 296 | |||
AL050215.1 | - | 2, 3 | 132 | |||
BC010130.2 | - | 4, 5 | 425 | 3953739 | ||
BP341272.1 | - | 4, 5 | 490 |
Gene Symbol: MGC72080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002822.3 | 2, 3 | 270 | |||
GenBank mRNA | BC029615.1 | - | 2, 3 | 280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
