siRNA Details
Gene Symbol: CCZ1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198097.3 | NP_932765.1 | 7 | 706 | ||
GenBank mRNA | AK292326.1 | - | 7 | 674 | ||
AK296026.1 | - | 6 | 512 | |||
AL050215.1 | - | 5 | 348 | |||
BC010130.2 | - | 7 | 641 | 3953739 |
Gene Symbol: CCZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015622.5 | NP_056437.4 | 7 | 698 | ||
GenBank mRNA | AF151801.1 | - | 7 | 742 | ||
BC067810.1 | - | 7 | 708 | |||
BC132749.1 | - | 7 | 683 | 40146771 | ||
BC132751.1 | - | 7 | 683 | 40146773 | ||
BI546694.1 | - | 7 | 689 | |||
CN310516.1 | - | 4 | 390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
