siRNA Details
Gene Symbol: CCZ1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198097.3 | NP_932765.1 | 12 | 1174 | ||
GenBank mRNA | AK292326.1 | - | 12 | 1142 | ||
AK296026.1 | - | 11 | 980 | |||
AL050215.1 | - | 10 | 816 | |||
BC010130.2 | - | 12 | 1109 | 3953739 |
Gene Symbol: CCZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015622.5 | NP_056437.4 | 12 | 1166 | ||
GenBank mRNA | AF151801.1 | - | 12 | 1210 | ||
AK090536.1 | - | 1 | 2213 | |||
BC067810.1 | - | 12 | 1176 | |||
BC132749.1 | - | 12 | 1151 | 40146771 | ||
BC132751.1 | - | 12 | 1151 | 40146773 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
