siRNA Details
Gene Symbol: GOLGA6L22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271664.2 | NP_001258593.2 | 9 | 1724 | ||
XM_011543882.1 | XP_011542184.1 | 9 | 1359 |
Gene Symbol: GOLGA6L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001413.3 | NP_001001413.3 | 8 | 1740 | ||
XM_017022085.1 | XP_016877574.1 | 8 | 1757 | |||
GenBank mRNA | AK097517.1 | - | 9 | 1785 | ||
BC127816.1 | - | 9 | 894 |
Gene Symbol: GOLGA6L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145004.2 | NP_001138476.2 | 8 | 1950 | ||
GenBank mRNA | AK093450.1 | - | 1 | 83 |
Gene Symbol: LOC100132202 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022770.1 | XP_016878259.1 | 9 | 1442 |
Gene Symbol: LOC102723623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720795.3 | XP_006720858.1 | 8 | 1759 | ||
XM_017022778.1 | XP_016878267.1 | 8 | 1763 |
Gene Symbol: LOC645202 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022779.1 | XP_016878268.1 | 9 | 1286 | ||
GenBank mRNA | AK302306.1 | - | 9 | 1262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
