siRNA Details
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 2 | 165 | ||
XM_011517658.2 | XP_011515960.1 | 2 | 282 | |||
XM_011517659.2 | XP_011515961.1 | 2 | 74 | |||
XM_011517660.2 | XP_011515962.1 | 2 | 74 | |||
XM_011517662.2 | XP_011515964.1 | 2 | 1578 | |||
XM_011517666.2 | XP_011515968.1 | 2 | 74 | |||
XM_011517671.2 | XP_011515973.2 | 2 | 282 | |||
XM_017014170.1 | XP_016869659.1 | 1 | 30 | |||
XM_017014171.1 | XP_016869660.1 | 2 | 104 | |||
XM_017014172.1 | XP_016869661.1 | 2 | 1682 | |||
XM_017014173.1 | XP_016869662.1 | 2 | 282 | |||
GenBank mRNA | AK024448.1 | - | 2 | 44 | ||
AK289708.1 | - | 2 | 168 |
Gene Symbol: LOC100134445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ786211.1 | - | 2 | 154 |
Gene Symbol: LOC100288778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130745.1 | 2 | 165 | |||
GenBank mRNA | CN404396.1 | - | 1 | 42 | ||
DA015248.1 | - | 2 | 51 | |||
DC319181.1 | - | 2 | 168 |
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024077.2 | 2 | 303 | |||
GenBank mRNA | AL137714.1 | - | 2 | 190 | ||
BX377099.2 | - | 2 | 55 | |||
BX537637.1 | - | 2 | 304 | |||
DA685507.1 | - | 2 | 451 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 2 | 856 | |||
GenBank mRNA | AK056232.1 | - | 2 | 697 | ||
AK057951.1 | - | 1 | 52 | |||
AK095183.1 | - | 2 | 50 | |||
AK290087.1 | - | 1 | 52 | |||
AK291582.1 | - | 2 | 65 | |||
AK295256.1 | - | 2 | 49 | |||
AK300161.1 | - | 2 | 51 | |||
BC048328.1 | - | 1 | 39 | |||
BC110996.1 | - | 2 | 541 | |||
BM839085.1 | - | 1 | 51 | |||
BQ722707.1 | - | 1 | 66 | |||
BX452679.2 | - | 176 |
Gene Symbol: WASH5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033266.1 | 2 | 66 | |||
GenBank mRNA | AK308635.1 | - | 2 | 76 |
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 2 | 77 | |||
GenBank mRNA | AK092583.1 | - | 2 | 51 | ||
AK296182.1 | - | 2 | 50 | |||
AK302089.1 | - | 2 | 122 | |||
AK308540.1 | - | 2 | 51 | |||
AK308566.1 | - | 2 | 51 | |||
AK309926.1 | - | 2 | 51 | |||
AK310121.1 | - | 2 | 51 | |||
AK310139.1 | - | 2 | 51 | |||
BC063470.1 | - | 2 | 53 | |||
DA054875.1 | - | 2 | 71 | |||
DA143319.1 | - | 2 | 77 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
