siRNA Details
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 2 | 190 | ||
XM_011517658.2 | XP_011515960.1 | 2 | 307 | |||
XM_011517659.2 | XP_011515961.1 | 2 | 99 | |||
XM_011517660.2 | XP_011515962.1 | 2 | 99 | |||
XM_011517662.2 | XP_011515964.1 | 2 | 1603 | |||
XM_011517666.2 | XP_011515968.1 | 2 | 99 | |||
XM_011517671.2 | XP_011515973.2 | 2 | 307 | |||
XM_017014170.1 | XP_016869659.1 | 1 | 55 | |||
XM_017014171.1 | XP_016869660.1 | 2 | 129 | |||
XM_017014172.1 | XP_016869661.1 | 2 | 1707 | |||
XM_017014173.1 | XP_016869662.1 | 2 | 307 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX452679.2 | - | 201 |
Gene Symbol: WASH5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033266.1 | 2 | 91 |
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 2 | 102 | |||
GenBank mRNA | AK092583.1 | - | 2 | 76 | ||
AK310121.1 | - | 2 | 76 | |||
BC063470.1 | - | 2 | 78 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
