siRNA Details
Gene Symbol: GOLGA8M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282468.1 | NP_001269397.1 | 13 | 1289 | ||
XM_005268284.4 | XP_005268341.1 | 13 | 2088 | |||
XM_006720654.3 | XP_006720717.1 | 13 | 1232 | |||
XM_011521951.2 | XP_011520253.1 | 14 | 1657 | |||
XM_011521952.2 | XP_011520254.1 | 13 | 2088 | |||
XM_011521953.2 | XP_011520255.1 | 13 | 2088 | |||
XM_011521954.2 | XP_011520256.1 | 13 | 1222 |
Gene Symbol: GOLGA8H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282490.1 | NP_001269419.1 | 13 | 1287 |
Gene Symbol: GOLGA8J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282472.1 | NP_001269401.1 | 13 | 1289 | ||
XM_006720651.3 | XP_006720714.1 | 13 | 1309 | |||
XM_011521948.2 | XP_011520250.1 | 13 | 1617 | |||
XM_011521950.2 | XP_011520252.1 | 13 | 1615 |
Gene Symbol: GOLGA8K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282493.1 | NP_001269422.1 | 13 | 1289 |
Gene Symbol: GOLGA8N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282494.1 | NP_001269423.1 | 13 | 1289 | ||
GenBank mRNA | BC101292.2 | - | 3 | 558 |
Gene Symbol: GOLGA8O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277308.1 | NP_001264237.1 | 13 | 1289 | ||
XM_011521989.2 | XP_011520291.1 | 13 | 1978 |
Gene Symbol: GOLGA8R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282484.1 | NP_001269413.1 | 13 | 1286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
