siRNA Details
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024077.2 | 9, 10 | 1457 | |||
GenBank mRNA | AK292759.1 | - | 8, 9 | 1064 | ||
AY217347.1 | - | 8, 9 | 1884 | |||
AY341936.1 | - | 4, 5 | 590 | |||
AY341937.1 | - | 4, 5 | 590 | |||
BC015400.1 | - | 1, 2 | 149 | |||
BC073913.1 | - | 4, 5 | 853 | |||
BC082240.1 | - | 6, 7 | 2152 | |||
BC093083.1 | - | 4, 5 | 866 | |||
BC106020.1 | - | 6, 7 | 894 | |||
BX537637.1 | - | 9, 10 | 1458 |
Gene Symbol: LOC100288778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130745.1 | 9, 10 | 1336 | |||
GenBank mRNA | AW881719.1 | - | 1, 2 | 513 | ||
BC063682.1 | - | 5, 6 | 682 |
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 9, 10 | 1336 | ||
XM_011517658.2 | XP_011515960.1 | 9, 10 | 1453 | |||
XM_011517659.2 | XP_011515961.1 | 9, 10 | 1235 | |||
XM_011517660.2 | XP_011515962.1 | 9, 10 | 1232 | |||
XM_011517662.2 | XP_011515964.1 | 9, 10 | 2739 | |||
XM_011517663.2 | XP_011515965.1 | 8, 9 | 2588 | |||
XM_011517664.2 | XP_011515966.1 | 8, 9 | 2585 | |||
XM_011517665.2 | XP_011515967.1 | 8, 9 | 1134 | |||
XM_011517666.2 | XP_011515968.1 | 8, 9 | 1133 | |||
XM_017014169.1 | XP_016869658.1 | 9, 10 | 1352 | |||
XM_017014170.1 | XP_016869659.1 | 9, 10 | 1415 | |||
XM_017014171.1 | XP_016869660.1 | 7, 8 | 1027 | |||
XM_017014172.1 | XP_016869661.1 | 7, 8 | 2605 | |||
GenBank mRNA | AK289708.1 | - | 9, 10 | 1348 | ||
BC094860.1 | - | 1, 2 | 377 | |||
CN404407.1 | - | 2, 3 | 233 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 9, 10 | 2010 | |||
GenBank mRNA | AK056232.1 | - | 9, 10 | 1855 | ||
AK057951.1 | - | 11, 12 | 1460 | |||
AK095183.1 | - | 8, 9 | 1456 | |||
AK290087.1 | - | 7, 8 | 1076 | |||
AK291582.1 | - | 8, 9 | 1471 | |||
AW604905.1 | - | 1, 2 | 102 | |||
BC048328.1 | - | 8, 9 | 1219 | |||
BC063555.1 | - | 8, 9 | 1645 | |||
BC110991.1 | - | 4, 5 | 619 | |||
BQ935819.1 | - | 4, 5 | 622 |
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 9, 10 | 1230 | |||
GenBank mRNA | BC094698.1 | - | 6, 7 | 2006 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
