siRNA Details
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 9 | 1182 | |||
GenBank mRNA | AK092583.1 | - | 7 | 1717 | ||
AK308566.1 | - | 7 | 920 | |||
BC063470.1 | - | 6 | 1096 | |||
BC094698.1 | - | 6 | 1958 |
Gene Symbol: LOC100288778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130745.1 | 9 | 1288 | |||
GenBank mRNA | AW881719.1 | - | 1 | 465 | ||
BC063682.1 | - | 5 | 634 |
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 9 | 1288 | ||
XM_011517658.2 | XP_011515960.1 | 9 | 1405 | |||
XM_011517659.2 | XP_011515961.1 | 9 | 1187 | |||
XM_011517660.2 | XP_011515962.1 | 9 | 1184 | |||
XM_011517662.2 | XP_011515964.1 | 9 | 2691 | |||
XM_011517663.2 | XP_011515965.1 | 8 | 2540 | |||
XM_011517664.2 | XP_011515966.1 | 8 | 2537 | |||
XM_011517665.2 | XP_011515967.1 | 8 | 1086 | |||
XM_011517666.2 | XP_011515968.1 | 8 | 1085 | |||
XM_017014169.1 | XP_016869658.1 | 9 | 1304 | |||
XM_017014170.1 | XP_016869659.1 | 9 | 1367 | |||
XM_017014171.1 | XP_016869660.1 | 7 | 979 | |||
XM_017014172.1 | XP_016869661.1 | 7 | 2557 | |||
GenBank mRNA | AK024481.1 | - | 2 | 2797 | ||
BC094860.1 | - | 1 | 329 | |||
CN404407.1 | - | 2 | 185 |
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024077.2 | 9 | 1409 | |||
GenBank mRNA | AK292759.1 | - | 8 | 1016 | ||
AL137714.1 | - | 7 | 2191 | |||
AY341936.1 | - | 4 | 542 | |||
AY341937.1 | - | 4 | 542 | |||
BC073913.1 | - | 4 | 805 | |||
BC082240.1 | - | 6 | 2104 | |||
BC093083.1 | - | 4 | 818 | |||
BX537637.1 | - | 9 | 1410 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 9 | 1962 | |||
GenBank mRNA | AK056232.1 | - | 9 | 1807 | ||
AK057951.1 | - | 11 | 1412 | |||
AK095183.1 | - | 8 | 1408 | |||
AK290087.1 | - | 7 | 1028 | |||
AK291582.1 | - | 8 | 1423 | |||
BC063555.1 | - | 8 | 1597 | |||
BC110991.1 | - | 4 | 571 |
Gene Symbol: WASH6P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC029274.1 | - | 2 | 197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
