siRNA Details
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 9 | 1311 | ||
XM_011517658.2 | XP_011515960.1 | 9 | 1428 | |||
XM_011517659.2 | XP_011515961.1 | 9 | 1210 | |||
XM_011517660.2 | XP_011515962.1 | 9 | 1207 | |||
XM_011517662.2 | XP_011515964.1 | 9 | 2714 | |||
XM_011517663.2 | XP_011515965.1 | 8 | 2563 | |||
XM_011517664.2 | XP_011515966.1 | 8 | 2560 | |||
XM_011517665.2 | XP_011515967.1 | 8 | 1109 | |||
XM_011517666.2 | XP_011515968.1 | 8 | 1108 | |||
XM_017014169.1 | XP_016869658.1 | 9 | 1327 | |||
XM_017014170.1 | XP_016869659.1 | 9 | 1390 | |||
XM_017014171.1 | XP_016869660.1 | 7 | 1002 | |||
XM_017014172.1 | XP_016869661.1 | 7 | 2580 | |||
GenBank mRNA | CN404407.1 | - | 2 | 208 |
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 9 | 1205 | |||
GenBank mRNA | AK092583.1 | - | 7 | 1740 | ||
BC094698.1 | - | 6 | 1981 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
