siRNA Details
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 10 | 2033 | |||
GenBank mRNA | AK056232.1 | - | 10 | 1878 | ||
AK057951.1 | - | 12 | 1483 | |||
AK095183.1 | - | 9 | 1479 | |||
AK097269.1 | - | 2 | 1259 | |||
AK290087.1 | - | 8 | 1099 | |||
AK291582.1 | - | 9 | 1494 | |||
AW604905.1 | - | 2 | 125 | |||
BC063555.1 | - | 9 | 1668 | |||
BC110991.1 | - | 5 | 642 |
Gene Symbol: LOC100288778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130745.1 | 10 | 1359 | |||
GenBank mRNA | AW274219.1 | - | 1 | 50 | ||
AW881719.1 | - | 2 | 536 | |||
BC063682.1 | - | 6 | 705 |
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 10 | 1359 | ||
XM_011517658.2 | XP_011515960.1 | 10 | 1476 | |||
XM_011517659.2 | XP_011515961.1 | 10 | 1258 | |||
XM_011517660.2 | XP_011515962.1 | 10 | 1255 | |||
XM_011517662.2 | XP_011515964.1 | 10 | 2762 | |||
XM_011517663.2 | XP_011515965.1 | 9 | 2611 | |||
XM_011517664.2 | XP_011515966.1 | 9 | 2608 | |||
XM_011517665.2 | XP_011515967.1 | 9 | 1157 | |||
XM_011517666.2 | XP_011515968.1 | 9 | 1156 | |||
XM_017014169.1 | XP_016869658.1 | 10 | 1375 | |||
XM_017014170.1 | XP_016869659.1 | 10 | 1438 | |||
XM_017014171.1 | XP_016869660.1 | 8 | 1050 | |||
XM_017014172.1 | XP_016869661.1 | 8 | 2628 | |||
GenBank mRNA | AK024481.1 | - | 2 | 3625 | ||
BC094860.1 | - | 2 | 400 | |||
CN404407.1 | - | 3 | 256 |
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024077.2 | 10 | 1480 | |||
GenBank mRNA | AK292759.1 | - | 9 | 1087 | ||
AL137714.1 | - | 7 | 3027 | |||
AY341936.1 | - | 5 | 613 | |||
AY341937.1 | - | 5 | 613 | |||
BC073913.1 | - | 5 | 876 | |||
BC082240.1 | - | 7 | 2175 | |||
BC093083.1 | - | 5 | 889 | |||
BX537637.1 | - | 10 | 1481 |
Gene Symbol: WASH6P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC029274.1 | - | 2 | 1027 |
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 10 | 1253 | |||
GenBank mRNA | AK092583.1 | - | 7 | 2545 | ||
BC094698.1 | - | 7 | 2029 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
