siRNA Details
Gene Symbol: FAM178B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122646.2 | NP_001116118.2 | 16 | 2160 | ||
NM_001172667.1 | NP_001166138.1 | 4 | 361 | |||
NM_016490.4 | NP_057574.2 | 4 | 479 | |||
XM_011511294.2 | XP_011509596.1 | 15 | 2088 | |||
XM_017004263.1 | XP_016859752.1 | 17 | 2325 | |||
XM_017004264.1 | XP_016859753.1 | 17 | 2286 | |||
XM_017004266.1 | XP_016859755.1 | 7 | 733 | |||
XM_017004267.1 | XP_016859756.1 | 4 | 338 | |||
XM_017004268.1 | XP_016859757.1 | 4 | 364 | |||
GenBank mRNA | AF151068.1 | - | 4 | 314 | ||
AK122705.1 | - | 10 | 1039 | |||
AK308698.1 | - | 10 | 1042 | |||
BC038583.1 | - | 4 | 440 | |||
BC039488.1 | - | 14 | 1427 | |||
BC051379.1 | - | 4 | 479 | |||
BX103404.1 | - | 4 | 339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
