siRNA Details
Gene Symbol: FAM178B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122646.2 | NP_001116118.2 | 17 | 2313 | ||
NM_001172667.1 | NP_001166138.1 | 5 | 514 | |||
NM_016490.4 | NP_057574.2 | 5 | 632 | |||
XM_011511294.2 | XP_011509596.1 | 16 | 2241 | |||
XM_017004263.1 | XP_016859752.1 | 18 | 2478 | |||
XM_017004264.1 | XP_016859753.1 | 18 | 2439 | |||
XM_017004266.1 | XP_016859755.1 | 8 | 886 | |||
XM_017004267.1 | XP_016859756.1 | 5 | 491 | |||
XM_017004268.1 | XP_016859757.1 | 5 | 517 | |||
GenBank mRNA | AJ710728.1 | - | 1 | 77 | ||
AK122705.1 | - | 11 | 1192 | |||
AK308698.1 | - | 11 | 1195 | |||
BC038583.1 | - | 5 | 593 | |||
BC039488.1 | - | 15 | 1580 | |||
BX103404.1 | - | 5 | 492 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
