siRNA Details
Gene Symbol: DEXI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014015.3 | NP_054734.2 | 1 | 610 | ||
XM_017023171.1 | XP_016878660.1 | 1 | 1217 | |||
GenBank mRNA | AF108145.2 | - | 1 | 226 | ||
AF129929.1 | - | 1 | 122 | |||
AK314111.1 | - | 1 | 579 | |||
BC001083.2 | - | 1 | 346 | 2958243 | ||
BC037569.1 | - | 1 | 610 | 5227606 | ||
BC052989.1 | - | 1 | 343 | 6194689 | ||
BC071683.1 | - | 1 | 509 | 5190179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
