siRNA Details
Gene Symbol: MAGEA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282501.1 | NP_001269430.1 | 5 | 1290 | ||
NM_001282502.1 | NP_001269431.1 | 4 | 1192 | |||
NM_001282504.1 | NP_001269433.1 | 3 | 1072 | |||
NM_001282505.1 | NP_001269434.1 | 2 | 1006 | |||
NM_005361.3 | NP_005352.1 | 4 | 1367 | |||
NM_175742.2 | NP_786884.1 | 6 | 1353 | |||
NM_175743.2 | NP_786885.1 | 6 | 1369 | |||
XM_017029519.1 | XP_016885008.1 | 5 | 1329 | |||
GenBank mRNA | AK290443.1 | - | 5 | 1290 | ||
BC013098.2 | - | 2 | 976 | 4343986 | ||
BC063681.1 | - | 4 | 1156 | 4403536 | ||
BC108720.1 | - | 4 | 1186 | 6065905 | ||
BC112158.1 | - | 1 | 874 | 8327626 | ||
BC112160.1 | - | 1 | 874 | 8327628 |
Gene Symbol: MAGEA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321400.1 | NP_001308329.1 | 6 | 1369 | ||
NM_001321401.1 | NP_001308330.1 | 6 | 1353 | |||
NM_001321402.1 | NP_001308331.1 | 5 | 1290 | |||
NM_001321403.1 | NP_001308332.1 | 4 | 1192 | |||
NM_001321404.1 | NP_001308333.1 | 2 | 1006 | |||
NM_153488.4 | NP_705692.1 | 3 | 1072 | |||
XM_017029406.1 | XP_016884895.1 | 5 | 1328 |
Gene Symbol: MAGEA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR541768.1 | - | 1 | 844 |
Gene Symbol: MAGEA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005363.3 | NP_005354.1 | 3 | 1051 | ||
NM_175868.2 | NP_787064.1 | 3 | 1115 | |||
GenBank mRNA | AK289437.1 | - | 3 | 1035 | ||
BC041599.1 | - | 3 | 1024 | 5458512 | ||
BC067731.1 | - | 3 | 1032 | 4828429 | ||
BX640600.1 | - | 3 | 1412 | |||
D32076.1 | - | 1 | 909 | |||
EU161101.1 | - | 1 | 844 | |||
U10339.1 | - | 1 | 844 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
