siRNA Details
Gene Symbol: MAGEA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011548.1 | NP_001011548.1 | 3 | 289 | ||
NM_001011549.1 | NP_001011549.1 | 3 | 286 | |||
NM_001011550.1 | NP_001011550.1 | 3 | 277 | |||
NM_002362.4 | NP_002353.3 | 3 | 292 | |||
XM_005274677.2 | XP_005274734.1 | 3 | 332 | |||
XM_005274678.3 | XP_005274735.1 | 3 | 290 | |||
XM_005274679.3 | XP_005274736.1 | 3 | 299 | |||
GenBank mRNA | AK300179.1 | - | 2 | 349 | ||
AK313862.1 | - | 3 | 328 | |||
AY954624.1 | - | 2 | 162 | |||
BC017723.1 | - | 3 | 289 | 4394783 | ||
BC096712.1 | - | 3 | 228 | |||
BM548926.1 | - | 3 | 286 | |||
BM562570.1 | - | 2 | 367 | |||
BX479311.1 | - | 3 | 289 | |||
CB120212.1 | - | 3 | 277 | |||
D32075.1 | - | 1 | 136 | |||
D32077.1 | - | 1 | 136 | |||
U10340.1 | - | 1 | 71 |
Gene Symbol: MAGEA10-MAGEA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204811.2 | NP_001191740.1 | 4 | 452 | ||
GenBank mRNA | BX109673.1 | - | 4 | 395 | ||
FY212575.1 | - | 5 | 471 |
Gene Symbol: MAGEA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282501.1 | NP_001269430.1 | 5 | 517 | ||
NM_001282502.1 | NP_001269431.1 | 4 | 419 | |||
NM_001282504.1 | NP_001269433.1 | 3 | 299 | |||
NM_001282505.1 | NP_001269434.1 | 2 | 233 | |||
NM_005361.3 | NP_005352.1 | 4 | 594 | |||
NM_175742.2 | NP_786884.1 | 6 | 580 | |||
NM_175743.2 | NP_786885.1 | 6 | 596 | |||
XM_017029519.1 | XP_016885008.1 | 5 | 556 | |||
GenBank mRNA | AK290443.1 | - | 5 | 517 | ||
AU100045.1 | - | 2 | 210 | |||
BC013098.2 | - | 2 | 203 | 4343986 | ||
BC063681.1 | - | 4 | 383 | 4403536 | ||
BC108720.1 | - | 4 | 413 | 6065905 | ||
BC109188.1 | - | 2 | 156 | |||
BC112158.1 | - | 1 | 101 | 8327626 | ||
BC112160.1 | - | 1 | 101 | 8327628 | ||
BG575302.1 | - | 6 | 541 | |||
BM781931.1 | - | 6 | 536 | |||
DR158683.1 | - | 3 | 284 |
Gene Symbol: MAGEA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321400.1 | NP_001308329.1 | 6 | 596 | ||
NM_001321401.1 | NP_001308330.1 | 6 | 580 | |||
NM_001321402.1 | NP_001308331.1 | 5 | 517 | |||
NM_001321403.1 | NP_001308332.1 | 4 | 419 | |||
NM_001321404.1 | NP_001308333.1 | 2 | 233 | |||
NM_153488.4 | NP_705692.1 | 3 | 299 | |||
XM_017029406.1 | XP_016884895.1 | 5 | 555 |
Gene Symbol: MAGEA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005362.3 | NP_005353.1 | 3 | 280 | ||
XM_005274676.3 | XP_005274733.1 | 3 | 354 | |||
XM_006724818.2 | XP_006724881.1 | 3 | 436 | |||
XM_011531160.2 | XP_011529462.1 | 3 | 260 | |||
XM_011531161.2 | XP_011529463.1 | 3 | 685 | |||
GenBank mRNA | AK223519.1 | - | 1 | 107 | ||
AK292384.1 | - | 3 | 280 | |||
BC000340.2 | - | 3 | 245 | 2822978 | ||
BC005963.1 | - | 3 | 280 | 4064879 | ||
BC011744.2 | - | 2 | 206 | 3345801 | ||
BC016803.1 | - | 3 | 242 | 4073332 | ||
BC017389.1 | - | 3 | 237 | 4047542 | ||
CR541767.1 | - | 1 | 71 | |||
CR541768.1 | - | 1 | 71 | |||
CR541774.1 | - | 1 | 71 | |||
CR542020.1 | - | 1 | 71 |
Gene Symbol: MAGEA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021049.4 | NP_066387.1 | 3 | 243 | ||
GenBank mRNA | AY310324.1 | - | 1 | 71 | ||
BC109187.1 | - | 3 | 243 | 40000127 |
Gene Symbol: MAGEA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005363.3 | NP_005354.1 | 3 | 278 | ||
NM_175868.2 | NP_787064.1 | 3 | 342 | |||
GenBank mRNA | AK289437.1 | - | 3 | 262 | ||
BC041599.1 | - | 3 | 251 | 5458512 | ||
BC067731.1 | - | 3 | 259 | 4828429 | ||
BE542284.1 | - | 3 | 233 | |||
BM015284.1 | - | 3 | 278 | |||
BM843807.1 | - | 3 | 330 | |||
BU182516.1 | - | 3 | 292 | |||
BX640600.1 | - | 3 | 639 | |||
D32076.1 | - | 1 | 136 | |||
EU161101.1 | - | 1 | 71 | |||
U10339.1 | - | 1 | 71 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
