siRNA Details
Gene Symbol: GNB1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053004.2 | NP_443730.1 | 2 | 153 | ||
GenBank mRNA | AB051432.1 | - | 2 | 94 | ||
AF238328.1 | - | 2 | 153 | |||
AF301895.1 | - | 2 | 118 | |||
AY007378.1 | - | 2 | 153 | |||
BC012060.1 | - | 2 | 54 | 4548339 |
Gene Symbol: C22orf29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024627.5 | NP_078903.3 | 2 | 222 | ||
GenBank mRNA | AK291262.1 | - | 2 | 190 | ||
AL832509.1 | - | 2 | 111 | |||
BX640785.1 | - | 1 | 311 | |||
BX640969.1 | - | 2 | 191 | |||
BX640998.1 | - | 2 | 191 | |||
DA182195.1 | - | 2 | 200 | |||
DA757523.1 | - | 2 | 190 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
