siRNA Details
Gene Symbol: ANKRD20A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012421.1 | NP_001012421.1 | 1 | 106 | ||
XM_011517877.2 | XP_011516179.1 | 1 | 106 | |||
XM_011517878.2 | XP_011516180.1 | 1 | 106 | |||
XM_011517879.2 | XP_011516181.1 | 1 | 56 |
Gene Symbol: ANKRD20A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032250.3 | NP_115626.2 | 1 | 106 | ||
XM_005272532.4 | XP_005272589.1 | 1 | 43 | |||
XM_011545682.2 | XP_011543984.1 | 1 | 43 | |||
GenBank mRNA | AL136793.1 | - | 1 | 107 |
Gene Symbol: ANKRD20A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012419.2 | NP_001012419.1 | 1 | 72 | ||
XM_005272525.3 | XP_005272582.1 | 1 | 328 | |||
XM_011543860.2 | XP_011542162.1 | 1 | 329 | |||
XM_011543861.2 | XP_011542163.1 | 1 | 333 |
Gene Symbol: ANKRD20A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098805.1 | NP_001092275.1 | 1 | 106 | ||
GenBank mRNA | AK307323.1 | - | 1 | 258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
