siRNA Details
Gene Symbol: MT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005953.3 | NP_005944.1 | 2, 3 | 187 | ||
GenBank mRNA | BC007034.1 | - | 1 | 172 | 4051220 | |
BC070289.1 | - | 1 | 166 | 4051053 | ||
BM746626.1 | - | 2, 3 | 187 | |||
BT007315.1 | - | 2, 3 | 97 | |||
M26637.1 | - | 2, 3 | 104 | |||
S52379.1 | - | 2, 3 | 97 | |||
V00594.1 | - | 2, 3 | 154 | |||
X97260.1 | - | 2, 3 | 97 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
