siRNA Details
Gene Symbol: MT1X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005952.3 | NP_005943.1 | 3 | 207 | ||
GenBank mRNA | BC018190.1 | - | 3 | 192 | 3859643 | |
BC032131.2 | - | 3 | 352 | 4053295 | ||
BC032338.1 | - | 3 | 194 | 5190391 | ||
CD101947.1 | - | 3 | 211 |
Gene Symbol: MT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005946.2 | NP_005937.2 | 3 | 209 | ||
GenBank mRNA | AF348995.1 | - | 3 | 191 | ||
AY028617.1 | - | 3 | 209 | |||
BC029475.1 | - | 3 | 206 | 4723818 |
Gene Symbol: MT1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301267.1 | NP_001288196.1 | 3 | 211 | ||
NM_005950.2 | NP_005941.1 | 3 | 208 | |||
GenBank mRNA | BC020757.1 | - | 3 | 211 | 4767886 | |
BC048334.1 | - | 1 | 1124 | |||
BM817643.1 | - | 3 | 211 | |||
CD175975.1 | - | 3 | 227 |
Gene Symbol: MT1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005951.2 | NP_005942.1 | 3 | 207 | ||
GenBank mRNA | BC008408.1 | - | 3 | 207 | 4274128 | |
BC059780.1 | - | 3 | 187 | 5227592 | ||
BC146930.1 | - | 3 | 158 | 9056858 | ||
BC146934.1 | - | 3 | 158 | 9056862 | ||
BT007171.1 | - | 3 | 136 | |||
X64177.1 | - | 3 | 159 |
Gene Symbol: MT1HL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276687.1 | NP_001263616.1 | 1 | 182 | ||
GenBank mRNA | AF333388.1 | - | 1 | 182 | ||
BC133030.1 | - | 1 | 176 | |||
BC133032.1 | - | 1 | 176 |
Gene Symbol: MT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005953.3 | NP_005944.1 | 3 | 226 | ||
GenBank mRNA | BC007034.1 | - | 1 | 211 | 4051220 | |
BC070289.1 | - | 1 | 205 | 4051053 | ||
BT007315.1 | - | 3 | 136 | |||
M26637.1 | - | 3 | 143 | |||
S52379.1 | - | 3 | 136 | |||
X97260.1 | - | 3 | 136 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
