siRNA Details
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 7 | 1675 | ||
NR_003494.1 | 6 | 1409 | ||||
XM_006716256.3 | XP_006716319.1 | 8 | 1770 | |||
XM_006716257.3 | XP_006716320.1 | 7 | 1840 | |||
XM_006716260.3 | XP_006716323.1 | 6 | 1584 | |||
XM_006716262.1 | XP_006716325.1 | 6 | 1427 | |||
XM_011543842.2 | XP_011542144.1 | 6 | 1849 | |||
XM_011543843.2 | XP_011542145.1 | 7 | 1788 | |||
XM_017013916.1 | XP_016869405.1 | 7 | 1816 | |||
XM_017013917.1 | XP_016869406.1 | 6 | 1737 | |||
XM_017013918.1 | XP_016869407.1 | 6 | 1709 | |||
XM_017013919.1 | XP_016869408.1 | 5 | 1298 | |||
GenBank mRNA | AK098138.1 | - | 7 | 1272 | ||
BC062615.1 | - | 7 | 1349 | |||
BC085016.1 | - | 8 | 1378 | |||
BC093808.1 | - | 1 | 224 | |||
BC093810.1 | - | 1 | 224 | |||
BC098427.1 | - | 7 | 1355 |
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273603.4 | XP_005273660.1 | 7 | 1705 | ||
XM_005273604.4 | XP_005273661.1 | 6 | 1624 | |||
XM_005273605.4 | XP_005273662.1 | 6 | 1525 | |||
XM_005273607.2 | XP_005273664.1 | 6 | 1563 | |||
XM_006716387.3 | XP_006716450.1 | 7 | 1726 | |||
XM_011544618.2 | XP_011542920.1 | 8 | 1777 | |||
XM_011544619.2 | XP_011542921.1 | 7 | 1744 | |||
XM_011544622.2 | XP_011542924.1 | 7 | 1646 | |||
XM_017013740.1 | XP_016869229.1 | 6 | 1707 | |||
XM_017013741.1 | XP_016869230.1 | 5 | 1644 | |||
XM_017013742.1 | XP_016869231.1 | 5 | 1626 |
Gene Symbol: FAM86DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024241.1 | 8 | 1767 | |||
GenBank mRNA | BC043342.2 | - | 7 | 1491 |
Gene Symbol: FAM86EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130740.1 | 4 | 1933 | |||
NR_130741.1 | 4 | 987 | ||||
NR_130742.1 | 4 | 937 |
Gene Symbol: FAM86HP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024252.1 | 5 | 1383 | |||
GenBank mRNA | AK124448.1 | - | 3 | 1151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
