siRNA Details
Gene Symbol: NPIPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282511.1 | NP_001269440.1 | 2 | 124 | ||
XM_005255738.4 | XP_005255795.1 | 2 | 246 | |||
XM_006720830.3 | XP_006720893.1 | 2 | 1144 | |||
XM_011522342.2 | XP_011520644.1 | 2 | 173 | |||
XM_017022841.1 | XP_016878330.1 | 3 | 197 | |||
XM_017022842.1 | XP_016878331.1 | 3 | 249 | |||
XM_017022843.1 | XP_016878332.1 | 3 | 310 | |||
XM_017022844.1 | XP_016878333.1 | 2 | 117 | |||
XM_017022845.1 | XP_016878334.1 | 3 | 197 | |||
XM_017022846.1 | XP_016878335.1 | 3 | 197 | |||
XM_017022847.1 | XP_016878336.1 | 2 | 137 | |||
GenBank mRNA | BQ101079.1 | - | 2 | 170 |
Gene Symbol: LOC105376752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522797.2 | XP_011521099.1 | 2 | 255 |
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006985.2 | NP_008916.2 | 2 | 124 | ||
XM_011522748.2 | XP_011521050.1 | 2 | 221 | |||
XM_011522749.2 | XP_011521051.1 | 3 | 333 | |||
XM_011522750.2 | XP_011521052.1 | 2 | 247 | |||
XM_011522751.1 | XP_011521053.1 | 2 | 220 | |||
XM_017023891.1 | XP_016879380.1 | 3 | 333 | |||
GenBank mRNA | AF132984.1 | - | 2 | 124 | ||
AK160377.1 | - | 21 | 4311 | |||
BE253379.1 | - | 3 | 392 | |||
KF175165.1 | - | 2 | 160 | |||
KF175166.1 | - | 2 | 160 | |||
KF175167.1 | - | 2 | 160 | |||
KF175169.1 | - | 2 | 124 | |||
KF175208.1 | - | 2 | 271 | |||
KF175209.1 | - | 3 | 342 | |||
KF175225.1 | - | 2 | 124 |
Gene Symbol: NPIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277324.1 | NP_001264253.1 | 2 | 124 | ||
XM_005255489.3 | XP_005255546.1 | 4 | 2543 | |||
XM_005255492.4 | XP_005255549.1 | 2 | 528 | |||
XM_006720917.3 | XP_006720980.2 | 4 | 2587 | |||
XM_011522587.1 | XP_011520889.1 | 4 | 2587 | |||
XM_011522593.1 | XP_011520895.1 | 4 | 2587 | |||
XM_011522594.2 | XP_011520896.1 | 4 | 2587 | |||
XM_011522595.1 | XP_011520897.1 | 4 | 2587 | |||
XM_011522596.1 | XP_011520898.1 | 2 | 151 | |||
XM_011522597.1 | XP_011520899.1 | 4 | 2587 |
Gene Symbol: NPIPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277323.1 | NP_001264252.1 | 2 | 124 | ||
XM_005255484.3 | XP_005255541.1 | 3 | 212 | |||
XM_005255485.4 | XP_005255542.1 | 2 | 528 | |||
XM_011522576.2 | XP_011520878.2 | 4 | 2577 | |||
XM_011522577.2 | XP_011520879.2 | 4 | 2498 | |||
XM_011522578.2 | XP_011520880.2 | 2 | 220 | |||
XM_011522580.2 | XP_011520882.2 | 2 | 151 | |||
XM_011522581.1 | XP_011520883.1 | 4 | 2587 | |||
XM_011522583.2 | XP_011520885.1 | 4 | 2587 | |||
XM_011522584.1 | XP_011520886.1 | 4 | 2587 | |||
XM_011522585.1 | XP_011520887.1 | 2 | 151 | |||
XM_011522586.1 | XP_011520888.1 | 4 | 2587 | |||
XM_017023553.1 | XP_016879042.1 | 4 | 2587 | |||
XM_017023554.1 | XP_016879043.1 | 3 | 2500 |
Gene Symbol: NPIPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277325.1 | NP_001264254.1 | 2 | 124 | ||
XM_011522329.2 | XP_011520631.1 | 4 | 535 | |||
XM_011522336.2 | XP_011520638.1 | 4 | 528 | |||
XM_011522337.1 | XP_011520639.1 | 2 | 151 | |||
GenBank mRNA | EL734608.1 | - | 2 | 124 | ||
KF175168.1 | - | 2 | 121 | |||
KF175170.1 | - | 2 | 124 | |||
KF175171.1 | - | 2 | 118 | |||
KF175172.1 | - | 2 | 124 |
Gene Symbol: NPIPA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282507.1 | NP_001269436.1 | 2 | 124 | ||
XM_005255735.4 | XP_005255792.1 | 2 | 246 | |||
XM_011522339.2 | XP_011520641.1 | 2 | 172 | |||
XM_017022833.1 | XP_016878322.1 | 3 | 197 | |||
XM_017022834.1 | XP_016878323.1 | 3 | 249 | |||
XM_017022835.1 | XP_016878324.1 | 3 | 310 | |||
XM_017022836.1 | XP_016878325.1 | 2 | 117 | |||
XM_017022837.1 | XP_016878326.1 | 3 | 197 | |||
XM_017022838.1 | XP_016878327.1 | 2 | 592 | |||
XM_017022839.1 | XP_016878328.1 | 3 | 197 | |||
XM_017022840.1 | XP_016878329.1 | 2 | 137 |
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036447.1 | 27 | 5429 | |||
GenBank mRNA | AK124516.1 | - | 22 | 3684 | ||
AK126798.1 | - | 22 | 3596 | |||
AK304604.1 | - | 7 | 1129 | |||
BC039707.1 | - | 3 | 443 | 4479933 | ||
BC046145.1 | - | 4 | 535 | |||
KF175174.1 | - | 2 | 160 | |||
KF175210.1 | - | 3 | 403 | |||
KF175211.1 | - | 4 | 475 | |||
KF175214.1 | - | 3 | 342 | |||
KF175219.1 | - | 2 | 310 |
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123721.1 | 20 | 4088 | |||
NR_123722.1 | 10 | 1949 | ||||
GenBank mRNA | AK131084.1 | - | 15 | 4452 | ||
AK131085.1 | - | 14 | 4358 | |||
AK302708.1 | - | 10 | 1949 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
