siRNA Details
Gene Symbol: NPIPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277323.1 | NP_001264252.1 | 5 | 510 | ||
XM_005255484.3 | XP_005255541.1 | 6 | 598 | |||
XM_005255485.4 | XP_005255542.1 | 5 | 914 | |||
XM_011522576.2 | XP_011520878.2 | 7 | 2963 | |||
XM_011522577.2 | XP_011520879.2 | 7 | 2884 | |||
XM_011522578.2 | XP_011520880.2 | 5 | 606 | |||
XM_011522580.2 | XP_011520882.2 | 5 | 537 | |||
XM_011522581.1 | XP_011520883.1 | 7 | 2973 | |||
XM_011522583.2 | XP_011520885.1 | 7 | 2973 | |||
XM_011522584.1 | XP_011520886.1 | 7 | 2973 | |||
XM_017023553.1 | XP_016879042.1 | 7 | 2973 | |||
XM_017023554.1 | XP_016879043.1 | 6 | 2886 |
Gene Symbol: NPIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277324.1 | NP_001264253.1 | 5 | 510 | ||
XM_005255489.3 | XP_005255546.1 | 7 | 2929 | |||
XM_005255492.4 | XP_005255549.1 | 5 | 914 | |||
XM_006720917.3 | XP_006720980.2 | 7 | 2973 | |||
XM_011522587.1 | XP_011520889.1 | 7 | 2973 | |||
XM_011522593.1 | XP_011520895.1 | 7 | 2973 | |||
XM_011522594.2 | XP_011520896.1 | 7 | 2973 | |||
XM_011522595.1 | XP_011520897.1 | 7 | 2973 |
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123721.1 | 24 | 4603 | |||
GenBank mRNA | AK131084.1 | - | 19 | 4967 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
