siRNA Details
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC094882.1 | - | 1 | 112 | 3093162 | |
KF175175.1 | - | 1 | 52 | |||
KF175179.1 | - | 1 | 52 | |||
KF175180.1 | - | 1 | 52 | |||
KF175182.1 | - | 1 | 52 | |||
KF175185.1 | - | 1 | 52 | |||
KF175187.1 | - | 1 | 52 | |||
KF175190.1 | - | 1 | 52 | |||
KF175201.1 | - | 1 | 52 | |||
KF175215.1 | - | 2 | 241 | |||
KF175224.1 | - | 1 | 52 |
Gene Symbol: LOC613037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023515.1 | XP_016879004.1 | 3 | 368 | ||
XM_017023516.1 | XP_016879005.1 | 3 | 637 | |||
XM_017023517.1 | XP_016879006.1 | 3 | 368 | |||
XM_017023518.1 | XP_016879007.1 | 3 | 456 | |||
XM_017023519.1 | XP_016879008.1 | 3 | 368 | |||
GenBank mRNA | BQ433543.1 | - | 225 |
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006985.2 | NP_008916.2 | 1 | 52 | ||
XM_011522749.2 | XP_011521051.1 | 2 | 261 | |||
XM_017023891.1 | XP_016879380.1 | 2 | 261 | |||
GenBank mRNA | AF132984.1 | - | 1 | 52 | ||
KF175208.1 | - | 1 | 53 | |||
KF175225.1 | - | 1 | 52 |
Gene Symbol: NPIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277324.1 | NP_001264253.1 | 1 | 52 | ||
XM_005255489.3 | XP_005255546.1 | 3 | 2471 | |||
XM_005255492.4 | XP_005255549.1 | 1 | 456 |
Gene Symbol: NPIPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277323.1 | NP_001264252.1 | 1 | 52 | ||
XM_005255484.3 | XP_005255541.1 | 2 | 140 | |||
XM_005255485.4 | XP_005255542.1 | 1 | 456 |
Gene Symbol: NPIPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277325.1 | NP_001264254.1 | 1 | 52 | ||
GenBank mRNA | EL734608.1 | - | 1 | 52 | ||
KF175168.1 | - | 1 | 49 | |||
KF175170.1 | - | 1 | 52 | |||
KF175172.1 | - | 1 | 52 |
Gene Symbol: NPIPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KF175204.1 | - | 1 | 52 |
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255741.3 | XP_005255798.1 | 1 | 333 | ||
XM_017023630.1 | XP_016879119.1 | 2 | 199 | |||
XM_017023631.1 | XP_016879120.1 | 1 | 333 |
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023626.1 | XP_016879115.1 | 2 | 226 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022820.1 | XP_016878309.1 | 2 | 208 | ||
XM_017022821.1 | XP_016878310.1 | 1 | 1074 | |||
XM_017022822.1 | XP_016878311.1 | 2 | 2077 | |||
XM_017022825.1 | XP_016878314.1 | 2 | 208 | |||
XM_017022826.1 | XP_016878315.1 | 2 | 208 |
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC039707.1 | - | 2 | 371 | 4479933 | |
BC046145.1 | - | 3 | 463 | |||
KF175210.1 | - | 1 | 53 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
