siRNA Details
Gene Symbol: LOC100996643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003846387.5 | XP_003846435.2 | 3 | 248 | ||
XM_005272561.4 | XP_005272618.1 | 3 | 248 | |||
XM_006716909.3 | XP_006716972.1 | 3 | 248 | |||
XM_006716910.3 | XP_006716973.1 | 2 | 209 | |||
XM_006716911.3 | XP_006716974.1 | 3 | 248 | |||
XM_006716912.3 | XP_006716975.1 | 3 | 255 | |||
XM_011545675.2 | XP_011543977.1 | 3 | 285 |
Gene Symbol: LOC100133920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024443.3 | 2 | 170 | |||
GenBank mRNA | AK074198.1 | - | 2 | 170 |
Gene Symbol: LOC101928195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135597.1 | 3 | 326 | |||
GenBank mRNA | CR740458.1 | - | 3 | 326 |
Gene Symbol: LOC101929583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136299.1 | 2 | 118 | |||
GenBank mRNA | DN831597.1 | - | 2 | 118 |
Gene Symbol: LOC102724580 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136300.1 | 2 | 394 |
Gene Symbol: LOC102724904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011518105.2 | XP_011516407.1 | 2 | 221 |
Gene Symbol: LOC105379443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545676.2 | XP_011543978.1 | 2 | 311 | ||
XM_011545677.2 | XP_011543979.1 | 2 | 311 | |||
XM_011545678.2 | XP_011543980.1 | 2 | 311 | |||
XM_017015396.1 | XP_016870885.1 | 2 | 272 |
Gene Symbol: LOC286297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046175.1 | 2 | 707 | |||
GenBank mRNA | AK097152.1 | - | 2 | 709 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
