siRNA Details
Gene Symbol: LOC728488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK296222.1 | - | 1 | 758 | ||
AK311627.1 | - | 1 | 218 |
Gene Symbol: LOC100652833 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543783.2 | XP_011542085.1 | 1 | 2211 |
Gene Symbol: LOC101929599 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010105.1 | XP_016865594.1 | 1 | 431 |
Gene Symbol: LOC102724728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136575.1 | 1 | 1640 | |||
GenBank mRNA | AK302597.1 | - | 1 | 1640 |
Gene Symbol: LOC107986351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010104.1 | XP_016865593.1 | 1 | 425 |
Gene Symbol: LOC441081 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073404.1 | 1 | 2357 | |||
GenBank mRNA | BI061609.1 | - | 1 | 310 | ||
DC422530.1 | - | 1 | 217 |
Gene Symbol: LOC727983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC112340.1 | - | 1 | 1377 |
Gene Symbol: POM121L10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024593.1 | 1 | 1640 |
Gene Symbol: POM121L4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024592.1 | 1 | 1732 | |||
GenBank mRNA | BC103891.2 | - | 1 | 1256 |
Gene Symbol: POM121L8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024583.1 | 1 | 1712 | |||
GenBank mRNA | AK302811.1 | - | 1 | 1715 | ||
AK308294.1 | - | 1 | 662 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
