siRNA Details
Gene Symbol: CA5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001739.1 | NP_001730.1 | 4 | 529 | ||
XM_011523309.2 | XP_011521611.1 | 4 | 598 | |||
XM_011523310.2 | XP_011521612.1 | 4 | 598 | |||
XM_017023646.1 | XP_016879135.1 | 4 | 598 | |||
GenBank mRNA | BC137405.1 | - | 4 | 525 | 9021403 | |
BC137411.1 | - | 4 | 525 | 9021409 | ||
L19297.1 | - | 4 | 528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
