siRNA Details
Gene Symbol: SPDYE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250742.4 | XP_005250799.1 | 3 | 578 | ||
XM_005250743.4 | XP_005250800.1 | 2 | 433 |
Gene Symbol: SPDYE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175064.2 | NP_778234.2 | 2 | 566 | ||
XM_005249719.3 | XP_005249776.1 | 4 | 1077 | |||
XM_017012015.1 | XP_016867504.1 | 4 | 1015 | |||
XM_017012016.1 | XP_016867505.1 | 3 | 427 | |||
GenBank mRNA | AF412027.1 | - | 2 | 575 | ||
AL137266.1 | - | 2 | 348 |
Gene Symbol: SPDYE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716211.3 | XP_006716274.1 | 2 | 707 | ||
XM_017012887.1 | XP_016868376.1 | 2 | 707 |
Gene Symbol: SPDYE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031618.3 | NP_001026789.2 | 4 | 1027 | ||
XM_011516237.2 | XP_011514539.1 | 4 | 1026 | |||
XM_017012223.1 | XP_016867712.1 | 4 | 1026 | |||
GenBank mRNA | AK302217.1 | - | 4 | 1027 | ||
BC100973.1 | - | 1 | 265 | 40005226 | ||
DR731150.1 | - | 1 | 171 | |||
JQ023160.1 | - | 3 | 553 |
Gene Symbol: SPDYE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166339.1 | NP_001159811.1 | 4 | 1027 | ||
XM_005250093.3 | XP_005250150.1 | 3 | 1595 | |||
XM_011515702.2 | XP_011514004.1 | 3 | 1595 |
Gene Symbol: SPDYE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306141.1 | NP_001293070.1 | 3 | 974 |
Gene Symbol: SPDYE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146210.2 | NP_001139682.2 | 3 | 974 |
Gene Symbol: SPDYE7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003666.2 | 1 | 265 | |||
GenBank mRNA | BC100974.2 | - | 1 | 265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
