siRNA Details
Gene Symbol: HBG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000184.2 | NP_000175.1 | 3 | 495 | ||
GenBank mRNA | AF110493.1 | - | 3 | 389 | ||
AK290492.1 | - | 3 | 495 | |||
BC010914.1 | - | 3 | 499 | 4134713 | ||
BC029387.1 | - | 3 | 525 | 4762402 | ||
BC130457.1 | - | 3 | 490 | 40146488 | ||
BC130459.1 | - | 3 | 490 | 40146490 | ||
X55656.1 | - | 3 | 432 | |||
X55657.1 | - | 1 | 78 |
Gene Symbol: HBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000559.2 | NP_000550.2 | 3 | 495 | ||
GenBank mRNA | BC010913.1 | - | 3 | 501 | 4133117 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
