siRNA Details
Gene Symbol: BAALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024372.1 | NP_001019543.1 | 1 | 158 | ||
NM_024812.2 | NP_079088.1 | 1 | 158 | |||
GenBank mRNA | AK022077.1 | - | 1 | 136 | ||
AK093819.1 | - | 11 | ||||
BC011517.1 | - | 1 | 100 | 4183341 | ||
BC035038.2 | - | 1 | 138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
