siRNA Details
Gene Symbol: BAALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024372.1 | NP_001019543.1 | 1 | 264 | ||
NM_024812.2 | NP_079088.1 | 1 | 264 | |||
GenBank mRNA | AF371319.1 | - | 1 | 59 | ||
AF371323.1 | - | 1 | 59 | |||
AK022077.1 | - | 1 | 242 | |||
AK093819.1 | - | 1 | 117 | |||
BC011517.1 | - | 1 | 206 | 4183341 | ||
BC035038.2 | - | 1 | 244 | |||
BI548687.1 | - | 1 | 265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
