siRNA Details
Gene Symbol: RPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000661.4 | NP_000652.2 | 3 | 197 | ||
NM_001024921.3 | NP_001020092.1 | 3 | 355 | |||
GenBank mRNA | AB062431.1 | - | 1 | 144 | ||
AK130966.1 | - | 1 | 160 | |||
AK297178.1 | - | 3 | 180 | |||
AK303908.1 | - | 1 | 160 | |||
AK312241.1 | - | 1 | 164 | |||
AL039355.2 | - | 3 | 330 | |||
AL832047.1 | - | 2 | 252 | |||
AY320412.1 | - | 3 | 180 | |||
AY376242.1 | - | 2 | 314 | |||
AY597415.1 | - | 1 | 171 | |||
BC000483.2 | - | 1 | 149 | 2964733 | ||
BC004156.2 | - | 1 | 143 | 2961026 | ||
BC004206.2 | - | 1 | 143 | 2961026 | ||
BC007967.2 | - | 1 | 199 | 4304670 | ||
BC012149.1 | - | 1 | 199 | 4554838 | ||
BC031906.1 | - | 2 | 144 | 4996986 | ||
BC066318.1 | - | 1 | 164 | 5277491 | ||
BC070214.1 | - | 1 | 203 | 6605055 | ||
BC100285.1 | - | 2 | 304 | 6022858 | ||
BG829769.1 | - | 2 | 145 | |||
BM841336.1 | - | 3 | 197 | |||
D14531.1 | - | 2 | 144 | |||
D16961.1 | - | 1 | 117 | |||
U09953.1 | - | 3 | 167 | |||
U21138.1 | - | 1 | 144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
