siRNA Details
Gene Symbol: RPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000661.4 | NP_000652.2 | 7 | 549 | ||
NM_001024921.3 | NP_001020092.1 | 7 | 707 | |||
GenBank mRNA | AB062431.1 | - | 1 | 496 | ||
AK130966.1 | - | 1 | 512 | |||
AK303908.1 | - | 2 | 453 | |||
AK312241.1 | - | 1 | 516 | |||
AL832047.1 | - | 3 | 3798 | |||
AY320412.1 | - | 7 | 532 | |||
AY376242.1 | - | 6 | 666 | |||
AY597415.1 | - | 1 | 523 | |||
BC000483.2 | - | 1 | 501 | 2964733 | ||
BC004156.2 | - | 1 | 495 | 2961026 | ||
BC004206.2 | - | 1 | 495 | 2961026 | ||
BC007967.2 | - | 1 | 551 | 4304670 | ||
BC012149.1 | - | 1 | 551 | 4554838 | ||
BC031906.1 | - | 6 | 496 | 4996986 | ||
BC066318.1 | - | 1 | 516 | 5277491 | ||
BC070214.1 | - | 1 | 555 | 6605055 | ||
BC100285.1 | - | 6 | 656 | 6022858 | ||
BG829769.1 | - | 6 | 497 | |||
D14531.1 | - | 6 | 496 | |||
U09953.1 | - | 7 | 519 | |||
U21138.1 | - | 1 | 496 |
Gene Symbol: RPL9P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130277.1 | - | 1 | 1126 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
