siRNA Details
Gene Symbol: NPIPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720830.3 | XP_006720893.1 | 1 | 1021 | ||
XM_017022842.1 | XP_016878331.1 | 2 | 126 | |||
XM_017022843.1 | XP_016878332.1 | 2 | 187 |
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522749.2 | XP_011521051.1 | 2 | 210 | ||
XM_017023891.1 | XP_016879380.1 | 2 | 210 |
Gene Symbol: NPIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255489.3 | XP_005255546.1 | 3 | 2420 | ||
XM_005255492.4 | XP_005255549.1 | 1 | 405 |
Gene Symbol: NPIPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255484.3 | XP_005255541.1 | 2 | 89 | ||
XM_005255485.4 | XP_005255542.1 | 1 | 405 |
Gene Symbol: NPIPA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022834.1 | XP_016878323.1 | 2 | 126 | ||
XM_017022835.1 | XP_016878324.1 | 2 | 187 | |||
XM_017022838.1 | XP_016878327.1 | 1 | 469 |
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255741.3 | XP_005255798.1 | 1 | 282 | ||
XM_017023630.1 | XP_016879119.1 | 2 | 148 | |||
XM_017023631.1 | XP_016879120.1 | 1 | 282 |
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023626.1 | XP_016879115.1 | 2 | 175 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022820.1 | XP_016878309.1 | 2 | 157 | ||
XM_017022821.1 | XP_016878310.1 | 1 | 1023 | |||
XM_017022822.1 | XP_016878311.1 | 2 | 2026 | |||
XM_017022825.1 | XP_016878314.1 | 2 | 157 | |||
XM_017022826.1 | XP_016878315.1 | 2 | 157 |
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC039707.1 | - | 2 | 320 | 4479933 | |
BC046145.1 | - | 3 | 412 | |||
KF175211.1 | - | 3 | 351 | |||
KF175212.1 | - | 4 | 283 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
