siRNA Details
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522749.2 | XP_011521051.1 | 1 | 65 | ||
XM_017023891.1 | XP_016879380.1 | 1 | 65 | |||
GenBank mRNA | AK160377.1 | - | 19 | 4089 | ||
BE253379.1 | - | 2 | 256 | |||
KF175209.1 | - | 2 | 206 |
Gene Symbol: NPIPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ101079.1 | - | 1 | 34 |
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036447.1 | 25 | 5207 | |||
GenBank mRNA | AK124516.1 | - | 20 | 3462 | ||
AK126798.1 | - | 20 | 3374 | |||
AK304604.1 | - | 6 | 993 | |||
BC039707.1 | - | 1 | 175 | 4479933 | ||
BC046145.1 | - | 2 | 267 | |||
KF175211.1 | - | 2 | 206 | |||
KF175214.1 | - | 2 | 206 | |||
KF175219.1 | - | 1 | 174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
