siRNA Details
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006985.2 | NP_008916.2 | 8 | 770 | ||
XM_011522748.2 | XP_011521050.1 | 8 | 867 | |||
XM_011522749.2 | XP_011521051.1 | 9 | 979 | |||
XM_011522750.2 | XP_011521052.1 | 7 | 785 | |||
XM_011522751.1 | XP_011521053.1 | 9 | 936 | |||
XM_017023891.1 | XP_016879380.1 | 10 | 1049 | |||
GenBank mRNA | AF132984.1 | - | 8 | 770 | ||
AK160377.1 | - | 28 | 5027 | |||
CR627265.1 | - | 8045 | ||||
KF175225.1 | - | 8 | 770 |
Gene Symbol: NPIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277324.1 | NP_001264253.1 | 8 | 770 | ||
XM_005255489.3 | XP_005255546.1 | 10 | 3189 |
Gene Symbol: NPIPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277323.1 | NP_001264252.1 | 8 | 770 | ||
XM_005255484.3 | XP_005255541.1 | 9 | 858 | |||
XM_011522576.2 | XP_011520878.2 | 10 | 3223 | |||
XM_011522577.2 | XP_011520879.2 | 10 | 3144 | |||
XM_011522578.2 | XP_011520880.2 | 8 | 866 | |||
XM_011522580.2 | XP_011520882.2 | 8 | 797 | |||
XM_017023553.1 | XP_016879042.1 | 10 | 3233 | |||
XM_017023554.1 | XP_016879043.1 | 9 | 3146 |
Gene Symbol: NPIPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277325.1 | NP_001264254.1 | 8 | 770 | ||
XM_011522336.2 | XP_011520638.1 | 11 | 1244 |
Gene Symbol: NPIPA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282507.1 | NP_001269436.1 | 8 | 770 | ||
XM_005255735.4 | XP_005255792.1 | 8 | 892 | |||
XM_011522339.2 | XP_011520641.1 | 8 | 818 | |||
XM_017022833.1 | XP_016878322.1 | 9 | 843 | |||
XM_017022834.1 | XP_016878323.1 | 9 | 895 | |||
XM_017022835.1 | XP_016878324.1 | 9 | 956 | |||
XM_017022836.1 | XP_016878325.1 | 8 | 763 | |||
XM_017022837.1 | XP_016878326.1 | 9 | 1706 | |||
XM_017022838.1 | XP_016878327.1 | 8 | 2101 | |||
XM_017022839.1 | XP_016878328.1 | 8 | 1598 | |||
GenBank mRNA | BI046418.1 | - | 5 | 421 | ||
BI832721.1 | - | 2 | 152 |
Gene Symbol: NPIPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282511.1 | NP_001269440.1 | 8 | 770 | ||
XM_005255738.4 | XP_005255795.1 | 8 | 892 | |||
XM_006720830.3 | XP_006720893.1 | 8 | 2653 | |||
XM_011522342.2 | XP_011520644.1 | 8 | 819 | |||
XM_017022841.1 | XP_016878330.1 | 9 | 843 | |||
XM_017022842.1 | XP_016878331.1 | 9 | 895 | |||
XM_017022843.1 | XP_016878332.1 | 9 | 956 | |||
XM_017022844.1 | XP_016878333.1 | 8 | 763 | |||
XM_017022845.1 | XP_016878334.1 | 9 | 1706 | |||
XM_017022846.1 | XP_016878335.1 | 8 | 1598 |
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036447.1 | 31 | 5906 | |||
GenBank mRNA | AF229069.1 | - | 1 | 129 | ||
AK124516.1 | - | 26 | 4161 | |||
BC039707.1 | - | 10 | 1159 | 4479933 | ||
BC046145.1 | - | 10 | 1181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
