siRNA Details
Gene Symbol: NXF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022053.3 | NP_071336.1 | 8 | 931 | ||
GenBank mRNA | AF285596.1 | - | 5 | 530 | ||
AJ277659.1 | - | 12 | 1417 | |||
AK000423.1 | - | 4 | 327 | |||
AK308255.1 | - | 8 | 870 | |||
BC015020.1 | - | 8 | 851 | 3921074 |
Gene Symbol: NXF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099686.2 | NP_001093156.1 | 8 | 931 |
Gene Symbol: NXF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032946.2 | NP_116564.2 | 6 | 644 | ||
NR_028089.1 | 6 | 644 | ||||
GenBank mRNA | AJ277654.1 | - | 6 | 644 | ||
AJ277655.1 | - | 6 | 644 | |||
AJ277656.1 | - | 6 | 644 | |||
AJ277657.1 | - | 4 | 455 | |||
AJ277658.1 | - | 4 | 455 | |||
BC131708.1 | - | 5 | 465 | 40121743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
