siRNA Details
Gene Symbol: IGKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF113887.1 | - | 2 | 77 | ||
BC070334.1 | - | 2 | 102 | |||
BC070335.1 | - | 2 | 103 | |||
BC070336.1 | - | 2 | 106 | |||
BC073763.1 | - | 2 | 91 | |||
BC073764.1 | - | 2 | 88 | |||
BC095489.1 | - | 2 | 97 | |||
BC110292.1 | - | 2 | 85 |
Gene Symbol: IGKV1-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC034146.1 | - | 2 | 102 |
Gene Symbol: SCFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | Y13056.1 | - | 407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
