siRNA Details
Gene Symbol: ZNF470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001668.3 | NP_001001668.3 | 6 | 2678 | ||
XM_017026801.1 | XP_016882290.1 | 7 | 2752 | |||
GenBank mRNA | AK129686.1 | - | 1 | 1442 | ||
AY484591.1 | - | 6 | 2194 | |||
BC136762.1 | - | 6 | 2190 | 9020752 | ||
BX641145.1 | - | 1 | 1641 |
Gene Symbol: LOC441666 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024380.1 | 3 | 1388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
