siRNA Details
Gene Symbol: PRAMEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294139.1 | NP_001281068.1 | 2 | 393 | ||
NM_023013.4 | NP_075389.2 | 4 | 1083 | |||
GenBank mRNA | AK125568.1 | - | 4 | 1083 | ||
AL049686.1 | - | 4 | 1083 | |||
BC040902.1 | - | 2 | 393 | |||
BC144199.1 | - | 4 | 1053 | 9052719 |
Gene Symbol: PRAMEF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291380.1 | NP_001278309.1 | 4 | 1083 | ||
XM_011541458.1 | XP_011539760.1 | 2 | 407 |
Gene Symbol: PRAMEF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024661.2 | NP_001019832.2 | 4 | 1083 | ||
XM_011542078.2 | XP_011540380.1 | 2 | 408 |
Gene Symbol: PRAMEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023014.1 | NP_075390.1 | 4 | 1067 | ||
XM_011542004.1 | XP_011540306.1 | 2 | 408 | |||
GenBank mRNA | AL049674.1 | - | 4 | 1067 | ||
BC075008.2 | - | 3 | 1016 | 30915615 | ||
BC075009.2 | - | 3 | 1016 | 30915402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
