siRNA Details
Gene Symbol: PRAMEF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024661.2 | NP_001019832.2 | 4 | 1663 | ||
XM_011542078.2 | XP_011540380.1 | 2 | 988 | |||
GenBank mRNA | BX092416.1 | - | 1 | 510 |
Gene Symbol: PRAMEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294139.1 | NP_001281068.1 | 2 | 973 | ||
NM_023013.4 | NP_075389.2 | 4 | 1663 | |||
GenBank mRNA | AK125568.1 | - | 4 | 1662 |
Gene Symbol: PRAMEF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291380.1 | NP_001278309.1 | 4 | 1663 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
