siRNA Details
Gene Symbol: PRAMEF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024661.2 | NP_001019832.2 | 4 | 1987 |
Gene Symbol: PRAMEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294139.1 | NP_001281068.1 | 2 | 1292 | ||
NM_023013.4 | NP_075389.2 | 4 | 1982 | |||
GenBank mRNA | AK125568.1 | - | 5 | 1955 | ||
BC040902.1 | - | 2 | 1292 |
Gene Symbol: PRAMEF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291380.1 | NP_001278309.1 | 4 | 1986 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
