siRNA Details
Gene Symbol: PRAMEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023014.1 | NP_075390.1 | 4 | 1566 | ||
XM_011542004.1 | XP_011540306.1 | 2 | 907 | |||
GenBank mRNA | AL049674.1 | - | 4 | 1566 | ||
DQ786308.1 | - | 1 | 357 |
Gene Symbol: PRAMEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294139.1 | NP_001281068.1 | 2 | 892 | ||
NM_023013.4 | NP_075389.2 | 4 | 1582 | |||
GenBank mRNA | AK125568.1 | - | 4 | 1581 | ||
AL049686.1 | - | 5 | 1582 | |||
BC040902.1 | - | 2 | 892 |
Gene Symbol: PRAMEF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291380.1 | NP_001278309.1 | 4 | 1582 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
