siRNA Details
Gene Symbol: PRAMEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294139.1 | NP_001281068.1 | 2 | 578 | ||
NM_023013.4 | NP_075389.2 | 4 | 1268 | |||
GenBank mRNA | AK125568.1 | - | 4 | 1268 | ||
AL049686.1 | - | 4 | 1268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
