siRNA Details
Gene Symbol: RPL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000983.3 | NP_000974.1 | 3, 4 | 295 | ||
GenBank mRNA | AK311749.1 | - | 3, 4 | 269 | ||
BC035566.1 | - | 3, 4 | 295 | 5109124 | ||
BC058887.1 | - | 3, 4 | 253 | 6141451 | ||
BC066314.1 | - | 3, 4 | 271 | 5303658 | ||
BU927988.1 | - | 309 | ||||
CA336134.1 | - | 3, 4 | 298 | |||
CR456873.1 | - | 3, 4 | 249 | |||
D17652.1 | - | 3, 4 | 271 | |||
X59357.1 | - | 2, 3 | 300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
