siRNA Details
Gene Symbol: RPL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000983.3 | NP_000974.1 | 3 | 190 | ||
GenBank mRNA | AK097075.1 | - | 3 | 166 | ||
AK311749.1 | - | 3 | 164 | |||
BC035566.1 | - | 3 | 190 | 5109124 | ||
BC058887.1 | - | 3 | 148 | 6141451 | ||
BC066314.1 | - | 3 | 166 | 5303658 | ||
BU927988.1 | - | 204 | ||||
CA336134.1 | - | 3 | 193 | |||
CR456873.1 | - | 3 | 144 | |||
D17652.1 | - | 3 | 166 | |||
X59357.1 | - | 2 | 195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
