siRNA Details
Gene Symbol: SLC29A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040661.1 | NP_001035751.1 | 7 | 807 | ||
NM_001300847.1 | NP_001287776.1 | 7 | 762 | |||
NM_153247.2 | NP_694979.2 | 7 | 804 | |||
XM_005249658.4 | XP_005249715.1 | 7 | 728 | |||
XM_006715667.3 | XP_006715730.1 | 7 | 675 | |||
XM_006715668.2 | XP_006715731.1 | 7 | 753 | |||
XM_011515200.2 | XP_011513502.1 | 7 | 835 | |||
XM_011515201.2 | XP_011513503.1 | 7 | 824 | |||
GenBank mRNA | AK075422.1 | - | 5 | 381 | ||
AK092242.1 | - | 7 | 790 | |||
AK296278.1 | - | 7 | 738 | |||
AY485959.1 | - | 7 | 804 | |||
BC025325.1 | - | 7 | 663 | 3354432 | ||
BC047592.1 | - | 7 | 768 | 4816109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
